Order Entry
Korea, Republic of
ContactUsLinkComponent
 
Irritant
Benzyl alcohol ≥99.5%, PESTINORM® for headspace gas chromatography
Catalog # 88250.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Benzyl alcohol ≥99.5%, PESTINORM® for headspace gas chromatography
Catalog # 88250.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Purity (GC, on anhydrous basis):
    99.5-100 %
  • Identity (IR):
    Complying
  • Residue after evaporation:
    0-0.0003 %w/w
  • Water (KF):
    0-0.1 %w/w
  • Color (APHA, Pt-Co):
    0-10
  • GC-HS (Each class 1 solvent):
    0-1 ug/g
  • GC-HS (Each class 2 solvent):
    0-10 ug/g
  • GC-HS (Each class 3 solvent):
    0-50 ug/g
  • Benzaldehyde content (on botteling):
    0-0.01 %
  • GC-HS (Each unknown):
    0-10 ug/g
  • Acidity:
    0-0.001 meq/gr
  • Alkalinity:
    0-0.002 meq/gr

Specification Test Results

About this item

 :
 :108.14 g/mol
Merck Index:13,01126
Boiling Pt:205 °C (1013 hPa)
Melting Pt:–15,3 °C
Density:1.05 g/cm³ (20 °C)
Flash Pt:101 °C
Storage Temperature:Ambient
EINECS:202-859-9
 :GHS07
CAS Number:100-51-6
Formula:C₆H₅CH₂OH
MDL Number:MFCD00004599