
Calcium chloride dihydrate 97-103%, granular, J.T.Baker®
5566-R
: Avantor
:
- Pk:300 lb
- Seq:7227
- USP - Assay (CaCl₂·2H₂O):99.0 - 107.0 %
- USP - Heavy Metals (as Pb):≤0.001 %
- USP - Identification:Passes Test
- USP - Iron, Aluminum, and Phosphate:Passes Test
- USP - Magnesium and Alkali Salts:≤1.0 %
- USP - pH of 5% Solution at 25°C:4.5 - 9.2
- EP/BP - Assay (CaCl₂·2H₂O):97.0 - 103.0 %
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Identification C:Passes Test
- EP/BP - Acidity or Alkalinity:Passes Test
- EP/BP - Sulfates (SO₄):≤300 ppm
- EP/BP - Aluminum (Al):Passes Test
- EP/BP - Barium (Ba):Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Heavy Metals (as Pb):≤20 ppm
- EP/BP - Iron (Fe):≤10 ppm
- EP/BP - Magnesium and Alkali Metals:≤0.5 %
:
:147.01 g/mol
:GHS07
:10035-04-8
:CaCl₂·2H₂O
:MFCD00149613