D-(-)-Sorbitol 91.0-100.5% (by anhydrous basis) NF, Multi-Compendial, J.T.Baker®
Catalog # 3449-X1
Supplier: Avantor
CAS Number:
Specifications
- Pk:1 KIT
- Seq:0075
Specifications
Specification Test Results
- Kit Components Meet Requirements:Passes Test
Specification Test Results
About this item
:
:182.17 g/mol
CAS Number:50-70-4
:98201-93-5
Formula:HOH₂C(CH(OH))₄CH₂OH
MDL Number:MFCD00004708