di-Potassium hydrogen phosphate kit 98.0-100.5% (dried basis) USP, J.T.Baker®
Catalog # 3251-X2
Supplier: Avantor
CAS Number:
Specifications
- Pk:1 KIT
- Seq:0075
Specifications
Specification Test Results
- Kit Components Meet Requirements:Passes Test
Specification Test Results
About this item
:
:174.18 g/mol
Merck Index:13,07743
Melting Pt:340 °C
Density:2.3 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-834-5
CAS Number:7758-11-4
Formula:K₂HPO₄
MDL Number:MFCD00011383