2-Propanol ≥99.9%, CMOS, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:67-63-0
:
Formula:(CH₃)₂CHOH
MDL Number:MFCD00011674
:60.1 g/mol
Merck Index:13,05228
Boiling Pt:82 °C (1013 hPa)
Melting Pt:–89 °C
Density:0.786 g/cm³ (20 °C)
Flash Pt:12 °C
Storage Temperature:Ambient
EINECS:200-661-7
UN:1219
ADR:3,II
REACH:01-2119457558-25
:GHS02
:GHS07
:GHS08
Specification Test Results
- Assay (CH₃CHOHCH₃):≥99.9 %
- Color (APHA):≤10
- Specific Gravity at 20°/20°C:0.785 - 0.787
- Residue after Evaporation:≤4 ppm
- Solubility in H₂O:Passes Test
- Water (H₂O)(by Karl Fischer titrn):≤0.05 %
- Acidity (µeq/g):≤0.2
- Alkalinity (µeq/g):≤0.1
- Bromide (Br):≤0.2 ppm
- Chloride (Cl):≤0.1 ppm
- Fluoride (F):≤0.200 ppm
- Nitrate (NO₃):≤0.2 ppm
- Nitrite (NO₂):≤0.2 ppm
- Phosphate (PO₄):≤0.3 ppm
- Sulfate (SO₄):≤0.2 ppm
- Trace Impurities - Aluminum (Al):≤50.0 ppb
- Arsenic and Antimony (as As):≤10.0 ppb
- Trace Impurities - Barium (Ba):≤20.0 ppb
- Trace Impurities - Beryllium (Be):≤100.0 ppb
- Trace Impurities - Bismuth (Bi):≤100.0 ppb
- Trace Impurities - Boron (B):≤10.0 ppb
- Trace Impurities - Cadmium (Cd):≤20.0 ppb
- Trace Impurities - Calcium (Ca):≤50.0 ppb
- Trace Impurities - Chromium (Cr):≤20.0 ppb
- Trace Impurities - Cobalt (Co):≤20 ppb
- Trace Impurities - Copper (Cu):≤10.0 ppb
- Trace Impurities - Gallium (Ga):≤50 ppb
- Trace Impurities - Germanium (Ge):≤50.0 ppb
- Trace Impurities - Gold (Au):≤20 ppb
- Heavy Metals (as Pb):≤200.0 ppb
- Trace Impurities - Iron (Fe):≤50.0 ppb
- Trace Impurities - Lead (Pb):≤20.0 ppb
- Trace Impurities - Lithium (Li):≤50.0 ppb
- Trace Impurities - Magnesium (Mg):≤20 ppb
- Trace Impurities - Manganese (Mn):≤15.0 ppb
- Trace Impurities - Molybdenum (Mo):≤100.0 ppb
- Trace Impurities - Nickel (Ni):≤10.0 ppb
- Trace Impurities - Niobium (Nb):≤100.0 ppb
- Trace Impurities - Potassium (K):≤100 ppb
- Trace Impurities - Silicon (Si):≤50 ppb
- Trace Impurities - Silver (Ag):≤20.0 ppb
- Trace Impurities - Sodium (Na):≤100.0 ppb
- Trace Impurities - Strontium (Sr):≤20.0 ppb
- Trace Impurities - Tantalum (Ta):≤100.0 ppb
- Trace Impurities - Thallium (Tl):≤10.0 ppb
- Trace Impurities - Tin (Sn):≤100 ppb
- Trace Impurities - Titanium (Ti):≤20 ppb
- Trace Impurities - Vanadium (V):≤100.0 ppb
- Trace Impurities - Zinc (Zn):≤50 ppb
- Trace Impurities - Zirconium (Zr):≤100.0 ppb
- Particle Count - 0.5 µm and greater:≤100 par/ml
- Particle Count - 1.0 µm and greater:≤10 par/ml
Specification Test Results
Frequently Bought Together