
meta-Phosphoric acid, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 0676.0500
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:79.98 g/mol
Boiling Pt:260 °C (1013 hPa)
Melting Pt:21 °C
Density:2.4 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:253-433-4
UN:3260
ADR:8,III
:GHS05
CAS Number:37267-86-0
Formula:(HPO₃)n
MDL Number:MFCD00011351