Titration solvent ASTM D-2710 / D-1159, VOLUSOL®
Catalog # 5203.2500
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
Specifications
Specification Test Results
- Density (20/4):1.05 - 1.07
- Identification:Passes test
Specification Test Results
About this item
:
UN:2920
ADR:8 (3),II
:GHS02
:GHS05
:GHS08
:GHS07
Storage Temperature:Ambient