Order Entry
Korea, Republic of
Orders LinkContactUsLinkComponent
Thiazine red, TECHNICAL

Thiazine red, TECHNICAL

Plastic bottle for solids, 10 g
Catalog # 27419.123
Irritant
Supplier:   VWR Chemicals
Thiazine red, TECHNICAL
Catalog # 27419.123
Supplier:  VWR Chemicals
CAS Number:  2150-33-6
Thiazine red, TECHNICAL
Thiazine red, TECHNICAL

Plastic bottle for solids, 10 g

CAS Number:  2150-33-6
Price
₩201,800
Log in for availability
1 * 10 g
Product Details & Documents
Synonyms:  Disodium 2-[4-[(1-hydroxy-4-sulphonato-2-naphthyl)azo]phenyl]-6-methylbenzothiazole-7-sulphonate
  • CAS Number:2150-33-6
  • Formula:H₂NCOCH₂NHCH₂CH₂SO₃H
  • Molecular Weight:599.58 g/mol
  • Storage Temperature:Ambient
  • EINECS:218-422-0

Documents

Certificates
Search by Lot Number
Specifications

Specification Test Results

  • Identification:
    Passes test

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    10 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.