

Liquid for manometer with sloping tube
Catalog # 24989.202
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:125 mL
- Seq:2475
Specifications
Specification Test Results
- Identification:Passes test
Specification Test Results
About this item
:
Flash Pt:14 °C (closed cup)
Storage Temperature:Ambient
UN:1170
ADR:3,II
:GHS02
:GHS07