Order Entry
Korea, Republic of
ContactUsLinkComponent
 
Oil of castor, GPR RECTAPUR®
 
undefined
Oil of castor, GPR RECTAPUR®
 

 

  • Solubility in methanol:
    Passes test
  • Acid value:
    Max. 1
  • Density (20/4):
    0.950 - 0.970
  • Iodine value:
    80 - 95
  • Saponification value:
    170 - 190
  • Non-saponifiable matter:
    Max. 0.8 %

 

Frequently Bought Together