

Glycolic acid 70% in water
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:79-14-1
:
Formula:C₂H₄O₃
MDL Number:MFCD00004312
:76.05 g/mol
Storage Temperature:Ambient
EINECS:201-180-5
UN:3265
ADR:8,II
:GHS05
:GHS08
Related Information
Certificates
Search by Lot NumberVWR Chemicals Big Packs and Bulks
Product Literature
VWRC85902.460_TDS_EN
Product specification
Frequently Bought Together