VWR®, Universal Pipette Tips
VWR® Universal low retention pipette tips repel liquid by creating a hydrophobic surface on the inside of the tip increasing your accuracy and data quality.
- Compatible with the widest range of pipette
- Maximises sample recovery with low binding resin and a fine point design
- Improves ergonomics by reducing the force required to attach and eject a tip
- Autoclavable at 15 psi, 122 °C for 15 minutes
- Manufactured under an ISO 9001 quality management system
VWR® tips use a low retention resin that is up to ten times more hydrophobic than other polypropylene pipette tips, enabling viscous samples like protein and DNA solutions to dispense easily without collecting inside the tip.
: Certified RNase-, DNase-, endotoxin-, metal-, and human DNA-free.
: Reloads will fit into emptied racks of respective racked tip version or into the empty racks 613-6445 or 613-6446, respectively.
VWR® Universal low retention pipette tips repel liquid by creating a hydrophobic surface on the inside of the tip increasing your accuracy and data quality.
- Compatible with the widest range of pipette
- Maximises sample recovery with low binding resin and a fine point design
- Improves ergonomics by reducing the force required to attach and eject a tip
- Autoclavable at 15 psi, 122 °C for 15 minutes
- Manufactured under an ISO 9001 quality management system
VWR® tips use a low retention resin that is up to ten times more hydrophobic than other polypropylene pipette tips, enabling viscous samples like protein and DNA solutions to dispense easily without collecting inside the tip.
: Certified RNase-, DNase-, endotoxin-, metal-, and human DNA-free.
: Reloads will fit into emptied racks of respective racked tip version or into the empty racks 613-6445 or 613-6446, respectively.
coc_VWRI613-6417_1010314306 coc_VWRI613-6418_1010263673 coc_VWRI613-6418_1010300082 coc_VWRI613-6421_1010069483 coc_VWRI613-6421_1010263679
- Catalog No:
- 613-6462
- 613-6453
- 613-6440
- 613-6433
- 613-6416
- 613-6463
- 613-6454
- 613-6441
- 613-6434
- 613-6417
- 613-6465
- 613-6456
- 613-6442
- 613-6435
- 613-6418
- 613-6468
- 613-6458
- 613-6436
- 613-6419
- 613-6469
- 613-6459
- 613-6443
- 613-6437
- 613-6420
- 613-6470
- 613-6460
- 613-6438
- 613-6421
- 613-6471
- 613-6461
- 613-6444
- 613-6439
- 613-6422
- Environmentally Preferable:
- Waste Reducing|Sustainable Packaging
- Waste Reducing|Sustainable Packaging
- Waste Reducing|Sustainable Packaging
- Waste Reducing|Sustainable Packaging
- Waste Reducing|Sustainable Packaging
- Waste Reducing|Sustainable Packaging
- Waste Reducing|Sustainable Packaging
- Waste Reducing|Sustainable Packaging
- Waste Reducing|Sustainable Packaging
- Waste Reducing|Sustainable Packaging
- Waste Reducing|Sustainable Packaging
- Waste Reducing|Sustainable Packaging
- Waste Reducing|Sustainable Packaging
- Waste Reducing|Sustainable Packaging
- Environmental certification:
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
Product Family Options
10 µl
- VolumeSterileLengthTip colourColour codePacked
Waste Reducing|Sustainable Packaging Specifications
Cat. No.613-6462Environmentally PreferableWaste Reducing|Sustainable PackagingEnvironmental certificationFSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plasticWaste Reducing|Sustainable Packaging Specifications
Cat. No.613-6453Environmentally PreferableWaste Reducing|Sustainable PackagingEnvironmental certificationFSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plasticWaste Reducing|Sustainable Packaging Specifications
Cat. No.613-6440Environmentally PreferableEnvironmental certificationSpecifications
Cat. No.613-6433Environmentally PreferableEnvironmental certificationSpecifications
Cat. No.613-6416Environmentally PreferableEnvironmental certification
10 µl extended
- VolumeSterileLengthTip colourColour codePacked
Waste Reducing|Sustainable Packaging Specifications
Cat. No.613-6463Environmentally PreferableWaste Reducing|Sustainable PackagingEnvironmental certificationFSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plasticWaste Reducing|Sustainable Packaging Waste Reducing|Sustainable Packaging Specifications
Cat. No.613-6454Environmentally PreferableWaste Reducing|Sustainable PackagingEnvironmental certificationFSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plasticSpecifications
Cat. No.613-6441Environmentally PreferableEnvironmental certificationSpecifications
Cat. No.613-6434Environmentally PreferableEnvironmental certificationSpecifications
Cat. No.613-6417Environmentally PreferableEnvironmental certification
200 µl
- VolumeSterileLengthTip colourColour codePacked
Specifications
Cat. No.613-6465Environmentally PreferableEnvironmental certificationWaste Reducing|Sustainable Packaging Specifications
Cat. No.613-6456Environmentally PreferableWaste Reducing|Sustainable PackagingEnvironmental certificationFSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plasticWaste Reducing|Sustainable Packaging Specifications
Cat. No.613-6442Environmentally PreferableWaste Reducing|Sustainable PackagingEnvironmental certificationFSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plasticSpecifications
Cat. No.613-6435Environmentally PreferableEnvironmental certificationSpecifications
Cat. No.613-6418Environmentally PreferableEnvironmental certification
250 µl
- VolumeSterileLengthTip colourColour codePacked
Waste Reducing|Sustainable Packaging Specifications
Cat. No.613-6468Environmentally PreferableWaste Reducing|Sustainable PackagingEnvironmental certificationFSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plasticWaste Reducing|Sustainable Packaging Waste Reducing|Sustainable Packaging Specifications
Cat. No.613-6458Environmentally PreferableWaste Reducing|Sustainable PackagingEnvironmental certificationFSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plasticSpecifications
Cat. No.613-6436Environmentally PreferableEnvironmental certificationSpecifications
Cat. No.613-6419Environmentally PreferableEnvironmental certification
300 µl
- VolumeSterileLengthTip colourColour codePacked
Specifications
Cat. No.613-6469Environmentally PreferableEnvironmental certificationWaste Reducing|Sustainable Packaging Specifications
Cat. No.613-6459Environmentally PreferableWaste Reducing|Sustainable PackagingEnvironmental certificationFSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plasticWaste Reducing|Sustainable Packaging Specifications
Cat. No.613-6443Environmentally PreferableWaste Reducing|Sustainable PackagingEnvironmental certificationFSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plasticWaste Reducing|Sustainable Packaging Specifications
Cat. No.613-6437Environmentally PreferableEnvironmental certificationSpecifications
Cat. No.613-6420Environmentally PreferableEnvironmental certification
1000 µl
- VolumeSterileLengthTip colourColour codePacked
Specifications
Cat. No.613-6470Environmentally PreferableEnvironmental certificationWaste Reducing|Sustainable Packaging Specifications
Cat. No.613-6460Environmentally PreferableWaste Reducing|Sustainable PackagingEnvironmental certificationFSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plasticSpecifications
Cat. No.613-6438Environmentally PreferableEnvironmental certificationWaste Reducing|Sustainable Packaging Specifications
Cat. No.613-6421Environmentally PreferableWaste Reducing|Sustainable PackagingEnvironmental certificationFSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
1250 µl
- VolumeSterileLengthTip colourColour codePacked
Specifications
Cat. No.613-6471Environmentally PreferableEnvironmental certificationWaste Reducing|Sustainable Packaging Specifications
Cat. No.613-6461Environmentally PreferableWaste Reducing|Sustainable PackagingEnvironmental certificationFSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plasticWaste Reducing|Sustainable Packaging Waste Reducing|Sustainable Packaging Specifications
Cat. No.613-6444Environmentally PreferableWaste Reducing|Sustainable PackagingEnvironmental certificationFSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plasticSpecifications
Cat. No.613-6439Environmentally PreferableEnvironmental certificationSpecifications
Cat. No.613-6422Environmentally PreferableEnvironmental certification
Empty racks
- ColourDescription
- TransparentEmpty racks for 1000 µl and 1250 µl tip reloads, universal and LTS® compatible, autoclavable1 * 10 items₩0