Glycerine ≥99.5%, AnalaR NORMAPUR® ACS, Reag. USP, double distilled
Catalog # BDH24388.320
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pk:2,5 L
- Seq:5096
Specifications
Specification Test Results
- Assay:Min. 99.5%
- Acrolein and Glucose:Passes Test
- Appearance of Solution (10 % V/V; water):Passes Test
- Substances Darkened by Sulphuric Acid:Passes Test
- Acidity:Max. 0.0004 meq/g
- Alkalinity:Max. 0.0003 meq/g
- Coloration:Max. 10 APHA
- Density (20/4):1.257 - 1.265
- Acetaldehyde:Max. 10 ppm
- Chlorinated Compounds (as Cl):Max. 0.003%
- Esters of Fatty Acids (as C₁₅H₂₆O₆):Max. 0.05%
- Glyceraldehyde:Max. 30 ppm
- Heavy Metals (as Pb):Max. 1 ppm
- Ignition Residue (SO₄):Max. 50 ppm
- Ketones (as CH₃COCH₃):Max. 50 ppm
- Water:Max. 0.5%
- Cl (Chloride):Max. 1 ppm
- NH₄ (Ammonium):Max. 5 ppm
- SO₄ (Sulphate):Max. 5 ppm
- As (Arsenic):Max. 1 ppm
- Ba (Barium):Max. 5 ppm
- Fe (Iron):Max. 1 ppm
- Conforms to ACS:Passes Test
- Conforms to Reag. USP:Passes Test
Specification Test Results
About this item
:
:92.09 g/mol
CAS Number:56-81-5
Formula:HOCH₂CH(OH)CH₂OH
MDL Number:MFCD00004722