Order Entry
Korea, Republic of
ContactUsLinkComponent
tri-Sodium citrate dihydrate USP, meets analytical specification of BP, Ph. Eur., JP, Macron Fine Chemicals™
tri-Sodium citrate dihydrate USP, meets analytical specification of BP, Ph. Eur., JP, Macron Fine Chemicals™
Catalog # MACR7855-06
CAS Number:  
undefined
tri-Sodium citrate dihydrate USP, meets analytical specification of BP, Ph. Eur., JP, Macron Fine Chemicals™
Catalog # MACR7855-06
CAS Number:  

Specifications

  • Pk:
    25 kg
  • Seq:
    4283

Specifications

About this item

 :
 :294.1 g/mol
CAS Number:6132-04-3
Formula:Na₃C₆H₅O₇·2H₂O
MDL Number:MFCD00150031