Order Entry
Korea, Republic of
Orders LinkContactUsLinkComponent
Rankem®, Büchner Funnels, Porcelain

Rankem®, Büchner Funnels, Porcelain

4800 ml, 275 mm, 240 mm
Catalog # RANKFB-4800-1
Supplier:   RANKEM
Rankem®, Büchner Funnels, Porcelain
Catalog # RANKFB-4800-1
Supplier:  RANKEM
Rankem®, Büchner Funnels, Porcelain

4800 ml, 275 mm, 240 mm

Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents

Made of white glazed porcelain.

Specifications
  • Capacity:
    4800 ml
  • Seq:
    09
  • Stem O-Ø:
    275 mm
  • For filter Ø:
    240 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.