


Perchloric acid 0.10 N in glacial acetic acid, StandARd® volumetric solution, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Certificates
About this item
CAS Number:7601-90-3
:
Formula:HClO₄
MDL Number:MFCD00011325
:100.46 g/mol
Storage Temperature:Ambient
EINECS:231-512-4
UN:2920
ADR:8 (3),II
:GHS02
:GHS05
:GHS07
Specification Test Results
- Normality:0.0995 - 0.1005
Specification Test Results
Frequently Bought Together