Order Entry
Korea, Republic of
ContactUsLinkComponent
VWR®, Cleanroom Socks, ESD, Disposable
VWR®, Cleanroom Socks, ESD, Disposable
Supplier:  VWR Collection
   Sustainable Options Available
Certificates
undefined
VWR®, Cleanroom Socks, ESD, Disposable
Supplier:  VWR Collection
   Sustainable Options Available
Specifications
  • Catalog No:
  • 113-9410
  • 113-9409
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Anti-static:
  • +
  • +
  • Cleanroom class:
  • 10/ISO 4
  • 10/ISO 4
  • Garment application:
  • Cleanroom, Medical, Laboratory, Clinical, Electronics/semi-conductor
  • Cleanroom, Medical, Laboratory, Clinical, Electronics/semi-conductor
  • Environmental certification:
  • ISO 14001 (Certificate registration No. 121311004)