VWR®, Multiwell Cell Culture Plates
Ideal for cell growth and cell yields.
- Uniform well volume
- Flat well bottom and round bottom plates available
- Well surface is smooth and free from striation
- Alphanumerically coded for easy identification
- Sterilised by gamma irradiation, non-pyrogenic
Raised rims on wells with the uniform rings on the lid help to reduce evaporation.
Plates are designed for use in applications where cell attachment is not desired. Plates are not autoclavable.
Single-position lid reduces the risk of cross-contamination and handling mistakes.
: ISO13485 2003 and ISO9001 2008 certified.
: Individually packaged in peel-to-open paper/plastic blister pack.
Frequently Bought Together