VWR® Shoe Covers, SMS
공급사: VWR Collection
인증서
이 제품에 관하여
These disposable sterile boot covers are manufactured from 60 g/m² spunbond meltblown spunbond SMS fabric and feature a 250 g/m² PVC sole.
- Bonded seams
- Elastic bands for a secure fit
- Slip resistant sole
- Sterility Assurance Level (SAL) of 10⁻⁶
Certifications: ISO 9001, ISO13485, ISO14001, GMP
Packaging: Boot covers are vacuum packed in pairs.
Frequently Bought Together