VWR®, Beakers, Low Form, Griffin
VWRU10754-948
: 10754-948
VWR® low form Griffin beakers are made from high quality Type 1, Class A, 33 expansion borosilicate glass. They feature a thick, slightly flared, beaded top with a spout designed to have excellent pouring characteristics.
- Double capacity scale
- Uniform wall thickness
- Very good chemical resistance
- High temperature resistance
Printed with white graduated scale and a durable marking area.
: Meets ASTM E960, E960-93.
Frequently Bought Together