Order Entry
Korea, Republic of
Orders LinkContactUsLinkComponent
VWR®, Micro cover glasses, rectangular
VWR®, Micro cover glasses, rectangular
Supplier:   VWR Collection
VWR®, Micro cover glasses, rectangular
Supplier:  VWR Collection
Certificates
VWR®, Micro cover glasses, rectangular
VWR®, Micro cover glasses, rectangular
12 Options Available Below
Product Details & Documents
These micro cover glasses are made of glass, and each set of numbered slides is uniform in size and thickness.
The No. 1 slides are 0,13 to 0,17 mm in thickness. The No. 1.5 slides are 0,16 to 0,19 mm in thickness.
Product Family Options
No. 1 slides
Product Information
  • L×W
    Availability
    Price
  • 22×30 mm
    Log in for availability
    Log in for availability
    1 * 28,35 g
    1 * 28,35 g - ₩23,818
    ₩23,818
    1 * 28,35 g
       ₩23,818
  • 22×50 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - ₩23,818
    ₩23,818
    1 * 1 items
       ₩23,818
  • 22×60 mm
    Log in for availability
    Log in for availability
    1 * 28,35 g
    1 * 28,35 g - ₩26,215
    ₩26,215
    1 * 28,35 g
       ₩26,215
  • 24×40 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - ₩23,782
    ₩23,782
    1 * 1 items
       ₩23,782
  • 24×50 mm
    Log in for availability
    Log in for availability
    1 * 1 oz
    1 * 1 oz - ₩22,097
    ₩22,097
    1 * 1 oz
       ₩22,097
  • 24×60 mm
    Log in for availability
    Log in for availability
    1 * 1 oz
    1 * 1 oz - ₩23,708
    ₩23,708
    1 * 1 oz
       ₩23,708
No. 1.5 slides
Product Information
  • L×W
    Availability
    Price
  • 22×40 mm
    Log in for availability
    Log in for availability
    1 * 28,35 g
    1 * 28,35 g - ₩23,444
    ₩23,444
    1 * 28,35 g
       ₩23,444
  • 22×50 mm
    Log in for availability
    Log in for availability
    1 * 1 oz
    1 * 1 oz - ₩23,669
    ₩23,669
    1 * 1 oz
       ₩23,669
  • 22×60 mm
    Log in for availability
    Log in for availability
    1 * 1 oz
    1 * 1 oz - ₩26,290
    ₩26,290
    1 * 1 oz
       ₩26,290
  • 24×40 mm
    Log in for availability
    Log in for availability
    1 * 28,35 g
    1 * 28,35 g - ₩23,556
    ₩23,556
    1 * 28,35 g
       ₩23,556
  • 24×50 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • 24×60 mm
    Log in for availability
    Log in for availability
    1 * 28,35 g
    1 * 28,35 g