

Acetonitrile with 0.1% (v/v) trifluoroacetic acid, BAKER ANALYZED® for LC-MS, J.T.Baker®
Supplier: Avantor
Certificates
About this item
:75-05-8
Storage Temperature:Ambient
EINECS:200-929-3
UN:1993
ADR:3,II
:GHS02
:GHS06
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:Passes Test
- Appearance:Passes Test
- Trifluoroacetic Acid:0.095 - 0.105 %
- Gradient Elution Test (a.u.) - 254 nm:≤0.01
- Residue after Evaporation:≤1.0 ppm
- Water (H₂O):≤200 ppm
- LC/MS Suitability - Largest Response on ESI- Positive Mode (as Reserpine):≤50 ppb
- Trace Impurities - Aluminum (Al):≤50.0 ppb
- Trace Impurities - * Calcium (Ca):≤50.0 ppb
- Trace Impurities - Iron (Fe):≤50.0 ppb
- Trace Impurities - Magnesium (Mg):≤50.0 ppb
- Trace Impurities - Potassium (K):≤50 ppb
- Trace Impurities - * Sodium (Na):≤50.0 ppb
Specification Test Results
Frequently Bought Together