Order Entry
Korea, Republic of
ContactUsLinkComponent
Calcium sulphate, Drierite™ 3 mm (8 mesh) for drying, without indicator, J.T.Baker®
Calcium sulphate, Drierite™ 3 mm (8 mesh) for drying, without indicator, J.T.Baker®
Catalog # L056-02
Supplier:  Avantor
CAS Number:  
Calcium sulphate, Drierite™ 3 mm (8 mesh) for drying, without indicator, J.T.Baker®
Catalog # L056-02
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Fibreboard box
  • Pk:
    2,3 kg
  • Seq:
    4070

Specifications

About this item

 :
 :136.14 g/mol
Merck Index:13,01711
Melting Pt:1350 °C
Density:2.964 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-900-3
CAS Number:7778-18-9
Formula:CaO₄S
MDL Number:MFCD00010912