J.T.Baker® BAKER INSTRA-ANALYZED®, Arsenic AA Standard
Catalog # 6442-04
Supplier: Avantor
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:5% HNO₃
- Pack type:Poly bottle for liquids, narrow mouth
- Pk:150 mL
- Element:Arsenic
- Symbol:As
Specifications
About this item
The standard is prepared by the careful dilution of the metal or its salt, usually 99,99% pure as verified by arc emission spectrometry
- Standardised with NIST standard chemicals
- Maximum shelf life and stability
The final concentration of each lot is determined by titrimetry or gravimetry.