
Mineral oil light, white NF, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Certificates
About this item
CAS Number:8012-95-1
:
MDL Number:MFCD00131611
Merck Index:13,07093
Boiling Pt:300…500 °C (1013 hPa)
Density:0.86 g/cm³ (20 °C)
Flash Pt:215 °C (closed cup)
EINECS:232-384-2
:GHS08
Specification Test Results
- Bulk Pharmaceutical Chemical:0.818 - 0.880
- NF - Specific Gravity at 25°/25°C:0.818 - 0.880
- NF - Viscosity at 40 C, mm₂/s:3.0 - 34.4
- NF - Readily Carbonizable Substances:Passes Test
- NF - Solid Paraffin:Passes Test
- NF - Acidity:Passes Test
- NF - Limit of Polycyclic Aromatic Hydrocarbons:Passes Test
- NF - Limit of Sulfur Compounds:Passes Test
- NF - Identification A:Passes Test
- NF - Identification B:Passes Test
Specification Test Results
Frequently Bought Together