Order Entry
Korea, Republic of
ContactUsLinkComponent
VWR®, Sealing Mats for Square Well Plates
VWR®, Sealing Mats for Square Well Plates
  VWRU76196-420
VWR®, Sealing Mats for Square Well Plates
  VWRU76196-420
  • Description:
    Sealing mat for 96 square well plate, sterile
  • Pk:
    50
Sealing mats made of EVA (Ethylene Vinyl Acetate), suitable for square well plates.
  • DNAse- and RNAse-free