
Amidosulphonic acid, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:5329-14-6
:
Formula:H₃NO₃S
MDL Number:MFCD00011603
:97.09 g/mol
Merck Index:13,09006
Boiling Pt:247 °C (1013 hPa)
Melting Pt:205 °C
Density:2.15 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:226-218-8
UN:2967
ADR:8,III
:GHS07
Specification Test Results
- Assay:min. 99.0 %
- Chloride (Cl):max. 0.001 %
- Heavy Metals (as Pb):max. 0.001 %
- Insoluble Matter:max. 0.005 %
- Residue after Ignition:max. 0.01 %
- Sulfate (SO4):max. 0.02 %
- Trace Impurities (in ppm) Iron (Fe):max. 5
Specification Test Results
Frequently Bought Together