Order Entry
Korea, Republic of
ContactUsLinkComponent
Ammonium sulphate ≥99.5%, Proteomics Grade
Irritant
Ammonium sulphate ≥99.5%, Proteomics Grade
Catalog # M105-1KG
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Ammonium sulphate ≥99.5%, Proteomics Grade
Catalog # M105-1KG
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    HDPE bottle
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Arsenic:
    <0.0001%
  • Chloride:
    <0.0005%
  • DNase:
    none detected
  • Heavy Metals (as Pb):
    <0.0002%
  • Insolubles:
    <0.005%
  • Iron:
    <0.0005%
  • Magnesium Test:
    Pass
  • pH (5%, water) @25 °C:
    5.0 – 6.0
  • Phosphate:
    <0.0002%
  • Protease:
    none
  • Purity (Dry Basis):
    >99.5%
  • Residue on Ignition:
    <0.005%
  • RNase:
    none detected
  • Selenium:
    ≤0.0005%

Specification Test Results

About this item

 :
 :132.14 g/mol
Merck Index:13,00559
Melting Pt:336…339 °C
Density:1.78 g/cm³ (20 °C)
EINECS:231-984-1
REACH:01-2119455044-46
 :GHS07
Storage Temperature:Ambient
CAS Number:7783-20-2
Formula:(NH₄)₂SO₄
MDL Number:MFCD00003391