

Sodium bisulphite ≥90% FCC, purified, Macron Fine Chemicals™
:7631-90-5
:
:HNaO₃S
:MFCD00167602
:12,08731
:300 °C
:2.476 g/cm³ (20 °C)
:231-548-0
:2693
:8,III
:104.06 g/mol
:GHS05
:GHS07
:Ambient
- Bulk Food Chemical:= 90 %
- FCC - Assay (as (Na₂S₂O₅)):≥90 %
- FCC - Assay (SO₂):58.5 - 67.4 %
- Solubility:Passes Test
- FCC - Identification A:Passes Test
- FCC - Identification B:Passes Test
- Insoluble Matter:≤0.005 %
- Thiosulfate (S₂O₃):≤0.05 %
- Arsenic (As):≤1 ppm
- FCC - Lead (Pb):≤2 mg/kg
- FCC - Iron (Fe):≤0.005 %
- FCC - Selenium (Se):≤5 mg/kg
- Chloride (Cl):≤0.02 %
Frequently Bought Together