
Potassium thiocyanate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Catalog # 27035.230
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:250 g
- Seq:3545
Specifications
Specification Test Results
- Assay:Min. 99.0 %
- pH (25°C; 5 %):5.3 - 8.5
- Heavy metals (as Pb):Max. 5 ppm
- Insolubility in water:Max. 30 ppm
- Substances reducing iodine (as I):Max. 0.025 %
- Cl (Chloride):Max. 50 ppm
- NH4 (Ammonium):Max. 20 ppm
- S (Sulphide):Max. 10 ppm
- SO4 (Sulphate):Max. 20 ppm
- Cu (Copper):Max. 2 ppm
- Fe (Iron):Max. 1 ppm
- Na (Sodium):Max. 50 ppm
- Pb (Lead):Max. 2 ppm
- Conforms to ACS:Passes test
- Conforms to Reag. Ph.Eur.:Passes test
Specification Test Results
About this item
:
Merck Index:12,07866
Boiling Pt:500 °C (1013 hPa)
Melting Pt:175 °C
Density:1.61 g/cm³ (20 °C)
:97.18 g/mol
Storage Temperature:Ambient
EINECS:206-370-1
:GHS07
CAS Number:333-20-0
Formula:KSCN
MDL Number:MFCD00011413