Order Entry
Korea, Republic of
ContactUsLinkComponent
Bromocresol green ACS
Bromocresol green ACS
Catalog # 0816-25G
Supplier:  VWR Chemicals
CAS Number:  
undefined
Bromocresol green ACS
Catalog # 0816-25G
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Clarity of Solution:
    Pass
  • Visual Transition Interval (pH 3.8 – 5.4):
    Pass

Specification Test Results

About this item

 :
 :698.02 g/mol
Merck Index:13,01370
Melting Pt:217…218 °C
Storage Temperature:Ambient
EINECS:200-972-8
CAS Number:76-60-8
Formula:C₂₁H₁₄Br₄O₅S
MDL Number:MFCD00005874