VWR® Basic, Overalls
Supplier: VWR Collection
Certificates
About this item
These disposable overalls, with collar, are made of non woven, spunbonded polypropylene (SPP), a lightweight fabric that provides protection in non hazardous, dirty work environments.
- Collar
- Zip closure (one direction)
- Elasticated wrists
- Free from natural rubber latex
Weight: 40 g/m²
Packaging: 1 piece per bag, 50 bags per case.
Specifications
- Catalog No:
- 113-9009
- 113-9008
- 113-9003
- 113-9002
- 113-9005
- 113-9004
- 113-9006
- 113-9007
- Garment application:
- General Purpose, Laboratory
- General Purpose, Laboratory
- General Purpose, Laboratory
- General Purpose, Laboratory
- General Purpose, Laboratory
- General Purpose, Laboratory
- General Purpose, Laboratory
- General Purpose, Laboratory
- Weight:
- 40 g/m²
- 40 g/m²
- 40 g/m²
- 40 g/m²
- 40 g/m²
- 40 g/m²
- 40 g/m²
- 40 g/m²