VWR® Basic, Overalls, Dual-Zip Closure
These disposable overalls, with collar, are made of non woven, spunbonded polypropylene (SPP), a lightweight fabric that provides protection in non hazardous, dirty work environments.
- Collar
- Zip closure (dual-direction)
- Elasticated wrists
- Free from natural rubber latex
Weight: 40 g/m²
: 1 piece per bag, 50 bags per case.
- Catalog No:
- 113-9011
- 113-9010
- 113-9013
- 113-9012
- Closure type:
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Garment application:
- General Purpose, Laboratory, Industrial
- General Purpose, Laboratory, Industrial
- General Purpose, Laboratory, Industrial
- General Purpose, Laboratory, Industrial
- Weight:
- 40 g/m²
- 40 g/m²
- 40 g/m²
- 40 g/m²
Frequently Bought Together