

Ammonia solution 10%, AnalaR NORMAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:1336-21-6
:7664-41-7
Formula:NH₃
MDL Number:MFCD00066650
:35.05 g/mol
Density:0.957 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-647-6
UN:2672
ADR:8,III
:GHS05
:GHS07
Specification Test Results
- Assay :Min. 10.0 %
- Appearance :Clear colourless liquid
- Cl (Chloride) :Max. 0.5 ppm
- PO₄ (Phosphate) :Max. 0.1 ppm
- S (Sulphide) :Max. 0.1 ppm
- SO₄ (Sulphate) :Max. 0.1 ppm
- Fe (Iron) :Max. 0.1 ppm
- K (Potassium) :Max. 1 ppm
- Mg (Magnesium) :Max. 0.5 ppm
- Mn (Manganese) :Max. 0.1 ppm
- Na (Sodium) :Max. 1 ppm
- Ni (Nickel) :Max. 0.05 ppm
- Pb (Lead) :Max. 0.05 ppm
- Zn (Zinc) :Max. 0.2 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101)
Packaging Information
spec_VWRC190063U spec_VWRC190063U spec_VWRC190063U spec_VWRC190064V spec_VWRC190064V
Product specification
More
Frequently Bought Together