Order Entry
Korea, Republic of
ContactUsLinkComponent
Chloroform ≥99.8% (by GC, corrected for water content) stabilised, BAKER ANALYZED® HPLC for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
자극성
건강
Chloroform ≥99.8% (by GC, corrected for water content) stabilised, BAKER ANALYZED® HPLC for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
공급사:  Avantor
인증서
undefined
자극성
건강
Chloroform ≥99.8% (by GC, corrected for water content) stabilised, BAKER ANALYZED® HPLC for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
공급사:  Avantor
인증서

사양 테스트 결과

  • For Laboratory,Research,or Manufacturing Use:
    = 0.01
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 400-280 nm:
    ≤0.01
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 254 nm:
    ≤0.15
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm:
    ≤245
  • Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission:
    ≤0.2 ppb
  • Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities:
    ≤1.0 ppb
  • Assay (CHCl₃) (by GC, exclusive of preservative, corrected for water):
    ≥99.8 %
  • Chloride (Cl):
    ≤10 ppm
  • Residue after Evaporation:
    ≤2.0 ppm
  • Substances Darkened by H₂SO₄:
    Passes Test
  • Titrable Acid (µeq/g):
    ≤0.5
  • Water (by KF, coulometric):
    ≤100 ppm

사양 테스트 결과

Frequently Bought Together