Order Entry
Korea, Republic of
ContactUsLinkComponent
VWR®, Cleanroom Wipes, 100% Polyester, Double Ply
VWR®, Cleanroom Wipes, 100% Polyester, Double Ply
Supplier:  VWR Collection
Certificates
undefined
VWR®, Cleanroom Wipes, 100% Polyester, Double Ply
Supplier:  VWR Collection
Certificates

Specifications

  • Catalog No:
  • 115-2470
  • 115-2446
  • 114-9323
  • Basis weight:
  • 220 g/m²
  • 220 g/m²
  • 220 g/m²

Specifications

Frequently Bought Together