VWR®, Transfer Pipettes for Blood Banks
These unbreakable all-in-one pipettes eliminate the hazard of broken glass and exposure to infectious materials. Molded from see-through low density PE, they are inert to biological fluids and most acids.
- Will not shatter or break
- Can be used in liquid nitrogen
- Non toxic and inert
- No bulb to insert or remove
- Uniform drop size
The low-affinity surface reduces the loss of cells and valuable proteins due to binding. Transfer pipettes can be sealed and refrigerated, and will work well whenever there is a need for quick, safe fluid transfer. Can be gas sterilised.
These unbreakable all-in-one pipettes eliminate the hazard of broken glass and exposure to infectious materials. Molded from see-through low density PE, they are inert to biological fluids and most acids.
- Will not shatter or break
- Can be used in liquid nitrogen
- Non toxic and inert
- No bulb to insert or remove
- Uniform drop size
The low-affinity surface reduces the loss of cells and valuable proteins due to binding. Transfer pipettes can be sealed and refrigerated, and will work well whenever there is a need for quick, safe fluid transfer. Can be gas sterilised.
Product Family Options
- CapacityDrops per mlBulb drawLengthSterilePacked