VWR®, Specimen containers, PP
VWRU89125-840
- Packed:Bulk
- Version:Sterile
- Capacity:148 ml
- Height:66,29 mm
- Pk:500
Sturdy, shatter-resistant containers manufactured from transparent polypropylene to allow constant sample viewing.
- Transparent siding
- Available in nonsterile or sterile formats
Containers feature molded graduations. Lids create a secure seal, and offer space for date, name, doctor, and room number molded into the surface.
: Polyethylene snap-on lids are sold separately.
: *The product(s) on this page are outside the scope of the EU Directive 2019/904