Order Entry
Korea, Republic of
ContactUsLinkComponent
Potassium hydrogen phthalate 99.95-100.05%, crystals, AR® ACS primary standard, Macron Fine Chemicals™
Potassium hydrogen phthalate 99.95-100.05%, crystals, AR® ACS primary standard, Macron Fine Chemicals™
Catalog # 670402.
CAS Number:  
undefined
Potassium hydrogen phthalate 99.95-100.05%, crystals, AR® ACS primary standard, Macron Fine Chemicals™
Catalog # 670402.
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, amber, wide mouth
  • Pk:
    125 g
  • Seq:
    3445

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    99.95 - 100.05 %
  • ACS - Assay (1-KOCOC₆H₄-2-COOH):
    99.95 - 100.05 %
  • ACS - Chlorine Compounds (as Cl):
    ≤0.003 %
  • ACS - Heavy Metals (as Pb):
    ≤5 ppm
  • ACS - Insoluble Matter:
    ≤0.005 %
  • ACS - Iron (Fe):
    ≤5 ppm
  • ACS - pH of 0.05M Solution at 25°C:
    4.00 - 4.02
  • ACS - Sodium (Na):
    ≤0.005 %
  • ACS - Sulfur Compounds (as S):
    ≤0.002 %

Specification Test Results

About this item

 :
 :204.22 g/mol
Merck Index:13,07694
Melting Pt:295…300 °C
Density:1.6362 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:212-889-4
CAS Number:877-24-7
Formula:HOOCC₆H₄COOK
MDL Number:MFCD00013070