Order Entry
Korea, Republic of
ContactUsLinkComponent
Ethanol denatured solution 70%, sterile made with water for injection (WFI) (denatured with methanol and isopropanol), J.T.Baker®
Flammable
Health
Irritant
Ethanol denatured solution 70%, sterile made with water for injection (WFI) (denatured with methanol and isopropanol), J.T.Baker®
Catalog # P004-03
Supplier:  Avantor
CAS Number:  
undefined
Flammable
Health
Irritant
Ethanol denatured solution 70%, sterile made with water for injection (WFI) (denatured with methanol and isopropanol), J.T.Baker®
Catalog # P004-03
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    1 gal US
  • Seq:
    8080

Specifications

Specification Test Results

  • CAUTION: For Manufacturing, processing or repackaging:
  • Appearance (Clear, colorless liquid):
    Passes Test
  • Specific Gravity:
    0.882 - 0.896
  • Nonvolatile Residue:
    Passes Test
  • Assay:
    68.0 - 72.0 %
  • Irradiation: Minimum Delivered Dose (K Gy):
    25.0 - 50.0
  • Irradiation: Maximum Delivered Dose (K Gy):
    25.0 - 50.0
  • Irradiation: Specified Dose Range (Lower):
    25.0 - 50.0
  • Irradiation: Specified Dose Range (Upper):
    25.0 - 50.0
  • Irradiation Date:
    25.0 - 50.0
  • Irradiation Batch Number:
  • The above referenced lot has passed the current USP Test for Sterility.:

Specification Test Results

About this item

 :
 :46.07 g/mol
ADR:3,III
 :Management of Change (MOC) category = NI
UN:1993
 :GHS02
 :GHS08
 :GHS07
CAS Number:64-17-5
Formula:H₃CCH₂OH
MDL Number:MFCD00003568