Order Entry
Korea, Republic of
ContactUsLinkComponent
 
VWR®, Digital Mini Block Heaters
  460-0334
   Waste Reducing
undefined
VWR®, Digital Mini Block Heaters
  460-0334
   Waste Reducing

 

  • Heating capacity:
    0,075 kW
  • No. of blocks:
    1
  • Temperature Range:
    Ambient +5...100 °C
  • Seq:
    001
  • Environmentally Preferable:
    Waste Reducing
  • W×D×H:
    127×160×123 mm
  • Weight:
    1,3 kg
  • Accuracy:
    ±0,5 °C
  • Pk:
    1
  • Temp. stability:
    ±0,2 °C
  • Type:
    Mini block heater**

 

 

These mini block heaters are ideal for applications that require accurate, repeatable results. These personal-sized units take up minimal bench space. Choice of model with or without heated lid. Units are ideal for immunoassays, ELISA, LAMP assays, enzyme reactions, denaturations and blood banking.

  • LED display for temperature and time
  • Temperature range from ambient 5 to 100 °C
  • Temperature accuracy: ±0,5 °C (models with heated lid: ±0,3 °C)
  • Microprocessor controlled with temperature calibration via external thermometer or temperature device
  • Timer range: 1 second to 160 hours, or continuous mode
  • Heated lid helps to reduce condensation within sample tube
  • Accommodates interchangeable mini heating blocks for tubes from 0,2 up to 50 ml capacity
 : Blocks for 15 and 50 ml tubes (460-0342 and 462-0343) cannot be used with the mini block heater with heated lid.
 : Heating blocks are not included, please order these separately.
 : ** EU/CH-plug *** UK-plug

Blocks