Order Entry
Korea, Republic of
ContactUsLinkComponent
VWR® London, Safety Spectacles
  111-1845
undefined
VWR® London, Safety Spectacles
  111-1845
  • Type:
    Anti-UV (2-1,2), anti-scratch, anti-fog
  • Colour:
    Light blue
  • Lenses:
    Clear PC
  • Pk:
    1
Stylish protective eyewear combining fashion, fit and function.
  • Moulded-in nose bridge
  • Soft, non slip rubber temple tips
  • Ideal for small facial contours
 : Compliant with EN 166, EN 170; VWR® EN 166 FT CE (frame); VWR® 2C-1,2 1 FT (lens)