Order Entry
Korea, Republic of
ContactUsLinkComponent
VWR® Stockholm, Safety Spectacles
VWR® Stockholm, Safety Spectacles
Catalog # 111-1844
공급사:  VWR Collection
undefined
VWR® Stockholm, Safety Spectacles
Catalog # 111-1844
공급사:  VWR Collection
사양
  • Type:
    Anti-UV, anti-scratch, anti-fog
  • Colour:
    Transparent/black
  • Description:
    Stockholm spectacles
  • Lenses:
    Clear PC
  • Pk:
    1
사양
이 제품에 관하여
Extremely lightweight, modern safety eyewear with outstanding field of vision and exceptional protection.
  • Moulded-in nose bridge provides uncompromised comfort and fit
  • Single-piece lens that will fit most faces
  • Soft rubber tip for additional comfort
Certifications: Compliant with EN 166:2001, EN 170, ISO 9001:2008; 166 FT CE (frame) 1 FT (lens)