Order Entry
Korea, Republic of
ContactUsLinkComponent
EDTA disodium salt dihydrate 0.1 M, BAKER ANALYZED® volumetric solution, J.T.Baker®
  5632-02
 :  Avantor
 :  
undefined
EDTA disodium salt dihydrate 0.1 M, BAKER ANALYZED® volumetric solution, J.T.Baker®
  5632-02
 :  Avantor
 :  
  • Pack type:
    PE bottle, narrow mouth
  • Pk:
    1 L
  • Seq:
    5037
  • For Laboratory,Research,or Manufacturing Use:
    0.098 - 0.102
  • Molarity (Mol/L):
    0.098 - 0.102
 :
 :372.24 g/mol
 :1.02 g/cm³
 :6381-92-6
 :C₁₀H₁₄N₂Na₂O₈·2H₂O
 :MFCD00150037