Order Entry
Korea, Republic of
ContactUsLinkComponent
Sodium hexanitritocobaltate(III), BAKER ANALYZED® ACS, J.T.Baker®
Oxidizing
Health
Irritant
Sodium hexanitritocobaltate(III), BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Oxidizing
Health
Irritant
Sodium hexanitritocobaltate(III), BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates
Specification Test Results
  • Meets ACS Reagent Chemical Requirements:
    ≤0.02%
  • For R&D use only. Not on TSCA Inventory.:
    Passes Test
  • Insoluble Matter:
    ≤0.02 %
  • Suitability for Potassium Determination:
    Passes Test