Order Entry
Korea, Republic of
ContactUsLinkComponent
 
VWR® Spinbar®, Magnetic stirring bars, PTFE, large cylindrical
  VWRU58948-974
undefined
VWR® Spinbar®, Magnetic stirring bars, PTFE, large cylindrical
  VWRU58948-974

 

  • Ø:
    27 mm
  • Type:
    Giant
  • Length:
    57 mm
  • Seq:
    02
  • Pk:
    10

 

 

Heavy duty PTFE coated stirring bar which is ideal for stirring large volumes of fluid in vessels on large magnetic stirrers or other large equipment.

  • Fits into vessels with neck diameter of 32 mm
  • Multifaceted surface adds turbulence relative to smooth cylindrical stirring bars of a similar size
  • Autoclave safe