VWR® Dust'it™, Blower
Catalog # VWRU36934-142
Supplier: VWR Collection
Specifications
- Description:Blower, Dust'it™
- Pk:1
Specifications
About this item
These blowers are ideal for delicate, difficult-to-clean, and inaccessible equipments. The nonconductive plastic sleeve tip prevents scratches for cleaning optical lenses, slides, circuit boards, and cuvets.
- Blows away dust, lint, and dirt
- Environmentally safe
- Powerful air-jet
- One-way inlet valve at the top keeps air from being drawn in