Order Entry
Korea, Republic of
ContactUsLinkComponent
VWR®, Absorbent Pads
VWR®, Absorbent Pads
Catalog # VWRU95057-860
Supplier:  VWR Collection
undefined
VWR®, Absorbent Pads
Catalog # VWRU95057-860
Supplier:  VWR Collection
Specifications
  • Colour:
    White
  • Description:
    Absorbent pads
  • L×W:
    305×305 mm
  • Pk:
    600
Specifications
About this item
Designed to soak up liquids, oils, and common solvents, absorbent pads provide an excellent alternative to paper towels and gauze.
  • Strong, highly absorbent, and lint-free
  • Efficient alternative to paper towels and gauze
Pads exhibit a cloth-like softness with no loose threads or fibers, making them ideal for use in coverslipping, slide preparation, and general laboratory cleanup.