Order Entry
Korea, Republic of
ContactUsLinkComponent
VWR® Talon®, Water bath clamps
VWR® Talon®, Water bath clamps
Catalog # VWRU21573-129
Supplier:  VWR Collection
undefined
VWR® Talon®, Water bath clamps
Catalog # VWRU21573-129
Supplier:  VWR Collection
Specifications
  • Spanning width:
    5 - 10 mm
  • Description:
    Water bath clamp, small, two pronged
  • Length:
    89 mm
  • Pk:
    1
Specifications
About this item
Hold a variety of apparatus, including glass tubes and thermometers, on glass water bath walls of varying thickness up to 9 mm (¹¹/₃₂"). Knurled thumbscrew tightens jaws to hold object firmly.