VWR®, Spatula, PTFE Coated
Catalog # VWRU82027-476
Supplier: VWR Collection
Specifications
- Length:152 mm
- Pk:1
Specifications
About this item
Stainless steel, PTFE coated, with wooden handle.
Sturdy, economical tools for handling, transferring, blending, or collecting chemical and biological samples. They can be used to transfer sticky or corrosive materials without scratching or damaging container surfaces.